Clips for ground joints, Clipox® NS
Leverandør: CLIPOX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10060 10010 10034 10029 10024 10019 10012 10045
201-0440EA 0 DKK
201-0440 201-0406 201-0437 201-0436 201-0435 201-0409 201-0407 201-0439
Clips for ground joints, Clipox® NS
Klemmer Klemmer til NS-slib
Stainless steel spring wire AISI 316 Ti.
  • Outstanding thermal, chemical and acid resistance
  • Easy to mount and to remove due to two-sided handles
  • Secure horizontal and vertical conjunction with vertical suspension in case of pressure overload
Certificeringer: According to ISO 9001