Blande-målecylindere, med prop, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216181403 216185405 216186307 216181703 216183601 216184409 216180801 216182408
215-3664EA 437 DKK
215-3664 215-3669 215-3670 215-3665 215-3667 215-3668 215-3663 215-3666
Blande-målecylindere, med prop, DURAN®
Cylindere Målecylindre
DURAN® borosilicate glass 3.3, clear, polyethylene stopper.
  • Standard ground joint
  • White graduations
  • USP standard
The large hexagonal base prevents the cylinder from rolling. The base is equipped with three knobs which increase its stability. the cylinders have uniform wall thickness over the entire measurement range, so wedge errors are avoided.
Certificeringer: ISO 4788