2,4-Dinitrobenzaldehyd ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
A15452.03 A15452.06 A15452.14
A15452.03EA 562 DKK
A15452.03 A15452.06 A15452.14
2,4-Dinitrobenzaldehyd ≥98%
2,4-Dinitrobenzaldehyde
Purity: 98% CAS No.: 528-75-6 Molecular Formula: C7H4N2O5 Formula Weight: 196.12 Melting Point: 65-68° Boiling Point: 190°/10mm Merck Reference: 14,3272 MDL No.: MFCD00013376
Formel: C₇H₄N₂O₅
Kogepunkt: 190 °C (10 mmHg)
Smeltepunkt: 65…68 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013376
CAS-nummer: 528-75-6
EINECS: 208-440-7
Merck Index: 14,3272