Cannon-Fenske viscometers for automatic measurements
Leverandør: SI Analytics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
285403712 285403729 285403745 285403753 285403761 285403778 285403786 285403794 285403807 285403737 285403815
SCOI285403712EA 3250 DKK
SCOI285403712 SCOI285403729 SCOI285403745 SCOI285403753 SCOI285403761 SCOI285403778 SCOI285403786 SCOI285403794 SCOI285403807 SCOI285403737 SCOI285403815
Cannon-Fenske viscometers for automatic measurements
Viskosimetre Kapillærviskosimetere
Cannon-Fenske viscometers for automatic measurements especially in routine analyses.
Suitable for transparent liquids, foaming liquids, and liquid mixture with highly volatile components.