Bell Jars, with knob, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
244605902 244606607 244606907
SCOT244605902EA 2620 DKK
SCOT244605902 SCOT244606607 SCOT244606907
Bell Jars, with knob, DURAN®
Dåser Glasklokker
DURAN® boroslicate glass 3.3, clear.
  • Good chemical and physical properties
  • Excellent thermal resistance
  • Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).