Flasker, smal åbning
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
15700 15701 15702 15703 15704
KART15700EA 22.2 DKK
KART15700 KART15701 KART15702 KART15703 KART15704
Flasker, smal åbning
Flasker Plastflasker
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
The cylindrical bottles are supplied with insert and cap. Thanks to the large neck they are suitable for powders, solid specimen, creamy or semisolid substances, crystals and hygroscopic substances.
Certificeringer: EC Regulation 1935/2004 • EC Regulation 10/2011