Distilling receivers, Bernauer
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05288101 05288001 05288002 05288004 05288102 05288104
201-0906EA 803 DKK
201-0906 LENZ05288001 LENZ05288002 LENZ05288004 LENZ05288102 LENZ05288104
Distilling receivers, Bernauer
Destillations komponenter Destillationsmodtagere
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.