Diagnostikflasker, smal hals, rund, uden skruelåg Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
362006-9050 362006-9025 362006-9125
NALG362006-9050EA 9490 DKK
NALG362006-9050 NALG362006-9025 NALG362006-9125
Diagnostikflasker, smal hals, rund, uden skruelåg Nalgene®
Flasker Plastflasker
Bottles without closure use in sample storage and the shipping of small volume, valuable reagents.
  • Nonsterile
  • Natural translucent PPCO provides excellent chemical resistance
  • Durable and feature uniform walls resistant to splitting or puncturing
  • Leakproof when assembled with closure
Certificeringer: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
Bestillingsoplysninger: Coloured closures are ordered seperately.