Diagnostikflasker, smal åbning, runde, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
362002-9050 362002-9125 362002-9025
NALG362002-9050EA 8760 DKK
NALG362002-9050 NALG362002-9125 NALG362002-9025
Diagnostikflasker, smal åbning, runde, Nalgene®
Flasker Plastflasker
Bottles without closure, used to store and ship small volumes of valuable reagents.
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof when assembled with Nalgene closure
Natural, translucent high-density polyethylene, durable, uniform walls resistant to splitting or puncturing, A sample product certificate providing cytotoxicity, USP Class VI, FDA compliance 21 CFR and other regulatory compliance information is available or can be requested from our certificates of compliance database.
Certificeringer: More Product Information