Aspiratorflasker, smal åbning, med stophane, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
247035402 247036304 247037309 247038605
SCOT247035402EA 1670 DKK
SCOT247035402 SCOT247036304 SCOT247037309 215-1008
Aspiratorflasker, smal åbning, med stophane, DURAN®
Flasker Aspirator
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High thermal resistance, autoclavable
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Neck with DIN thread GL 45, tubulator with GL 32. Complete with screw connection cap, silicone seal and also stopcock with PTFE spindle. Dosing of liquids is possible via stopcock.
With printed retrace code (batch identification).