Diagnostikflasker, smal åbning, runde, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
362008-9125 362008-9025 362008-9050
NALG362008-9125EA 13650 DKK
NALG362008-9125 NALG362008-9025 NALG362008-9050
Diagnostikflasker, smal åbning, runde, Nalgene®
Flasker Plastflasker
Opaque bottles without closure, used to store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples.
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof when assembled with Nalgene closure
Certificeringer: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
Bestillingsoplysninger: Coloured closures are ordered seperately.