Vacuum hand pumps
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1398 1399
KART1398EA 2670 DKK
KART1398 KART1399
Vacuum hand pumps
Pumper Vakuumhåndpumper
Vacuum pumps have built in vacuum release valve allows for quick and easy release of the vacuum without disconnecting the pump.
  • Light and portable hand operated pumps
  • Easily dismantled for cleaning
  • Nozzle fits standard tubing of Int.Ø 6 mm
The release valve cap is transferred to the vacuum port to use as a pressure pump, enabling this pumps to be used for varied applications such as the transfer of liquids between vessels.