3-Bromanisol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A15625.14
A15625.22
A15625.36
A15625.14EA
498
DKK
A15625.14
A15625.22
A15625.36
3-Bromanisol ≥98%
3-Bromanisol
Purity: 98+%
CAS No.: 2398-37-0
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Flash Point: 93°(199°F)
Refractive Index: 1.5640
Melting Point: ca 2°
Boiling Point: 210-211°
Beilstein Reference: 1524687
MDL No.: MFCD00000081
Formel:
C₇H₇BrO Kogepunkt: 210…211 °C Smeltepunkt: ∼2 °C Massefylde: 1,489 Flammepunkt: 93 °C (199 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000081 CAS-nummer: 2398-37-0 EINECS: 219-264-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...