Toluen-p-sulfonicanhydrid ≥97%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride
L00160.06 L00160.14
L00160.06EA 659 DKK
L00160.06 L00160.14
Toluen-p-sulfonicanhydrid ≥97%
Toluen-p-sulfonicanhydrid
Purity: 97% CAS No.: 4124-41-8 Molecular Formula: C14H14O5S2 Formula Weight: 326.38 Melting Point: 125-130° Beilstein Reference: 2223702 UN No.: 3261 MDL No.: MFCD00008548
Formel: C₁₄H₁₄O₅S₂
Molekylvægt: 326.39
Smeltepunkt: 125…130 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008548
CAS-nummer: 4124-41-8
EINECS: 223-926-9
UN nummer: 3261
ADR: 8 III