2-Hydroxyethylmethacrylat ≥97% stabilised
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
HEMA, GMA, Methacrylic acid 2-hydroxyethyl ester, 1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate
Stabilization:
stabiliseret med hydroquinon monomethyl ether
Stabilized with 4-methoxyphenol
Purity: 97%
CAS No.: 868-77-9
Molecular Formula: CH2=CH(CH3)CO2CH2CH2OH
Formula Weight: 130.14
Storage Temperature: 2°C to 8°C
Flash Point: 101°(213°F)
Refractive Index: 1.4530
Odour: Ester-like
Melting Point: -12°
Boiling Point: 250°
Beilstein Reference: 1071583
MDL No.: MFCD00002863
Formel:
C₆H₁₀O₃ Kogepunkt: 250 °C Smeltepunkt: –12 °C Massefylde: 1,071 Flammepunkt: 101 °C (213 °F) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00002863 CAS-nummer: 868-77-9 EINECS: 212-782-2 |