1,1',1''-Nitrilotripropan-2-ol 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1',1''-Nitrilotripropan-2-ol, 1,1',1"-Nitrilotri-2-propanol, Tris(2-hydroxypropyl)amine

A19966.36 A19966.0E A19966.0C
A19966.36EA 292 DKK
A19966.36 A19966.0E A19966.0C
1,1',1''-Nitrilotripropan-2-ol 95%
1,1',1''-Nitrilotripropan-2-ol

Purity: 95% CAS No.: 122-20-3 Molecular Formula: C9H21NO3 Formula Weight: 191.27 Flash Point: 110°(230°F) Odour: Amine-like Boiling Point: 189-191°/23mm Beilstein Reference: 1071570 MDL No.: MFCD00004533

Formel: C₉H₂₁NO₃
Molekylvægt: 191.27 g/mol
Kogepunkt: 189…191 °C (23 mmHg)
Smeltepunkt: 48…52 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004533
CAS-nummer: 122-20-3
EINECS: 204-528-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR