Dapson 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4'-Diaminodiphenylsulfon
A18976.22
A18976.36
A18976.22EA
578
DKK
A18976.22
A18976.36
Dapson 98%
Dapson
Purity: 98%
CAS No.: 80-08-0
Molecular Formula: C12H12N2O2S
Formula Weight: 248.30
Odour: Odourless
Melting Point: 175-178°
Beilstein Reference: 788055
Merck Reference: 14,2822
MDL No.: MFCD00007887
Formel:
C₁₂H₁₂N₂O₂S Molekylvægt: 248.31 g/mol Smeltepunkt: 175…178 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007887 CAS-nummer: 80-08-0 EINECS: 201-248-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...