Isopropyl trifluoracetat ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopropyltrifluoroacetate, Trifluoroacetic isopropyl ester

A17322.14 A17322.22
A17322.14EA 477 DKK
A17322.14 A17322.22
Isopropyl trifluoracetat ≥97%
Isopropyl trifluoracetat

Purity: 97% CAS No.: 400-38-4 Molecular Formula: CF3CO2CH(CH3)2 Formula Weight: 156.10 Flash Point: -8°(17°F) Refractive Index: 1.3190 Melting Point: -78° Boiling Point: 73° Beilstein Reference: 1764312 UN No.: 2924 MDL No.: MFCD00013566

Formel: C₅H₇F₃O₂
Molekylvægt: 156.1 g/mol
Kogepunkt: 73 °C
Smeltepunkt: < –78 °C
Massefylde: 1.108
Flammepunkt: –8 °C (17 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013566
CAS-nummer: 400-38-4
EINECS: 206-922-1
UN nummer: 2924
ADR: 3 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR