Fluoranthen 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

119170250 119171000 119175000
ACRO119170250EA 375 DKK
ACRO119170250 ACRO119171000 ACRO119175000
Fluoranthen 98%
Fluoranthene
Formel: C₁₆H₁₀
Molekylvægt: 202.26 g/mol
Kogepunkt: 384 °C (760 mmHg)
Smeltepunkt: 109…111 °C
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001184
CAS-nummer: 206-44-0
EINECS: 205-912-4
UN nummer: 3077
ADR: 9 III

Bestil nu

Specifikationer Testresultater

Appearance Yellow to yellow-green to beige to light grey Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 106 to 111 °C
GC ≥97.5 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR