Flasker, bred hals, PP, rund, med skruelåg
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101789 101589 101689 101889 101989 102089 102189
215-2980EA 499 DKK
215-2980 215-2978 215-2979 215-2981 215-2982 VITL102089 VITL102189
Flasker, bred hals, PP, rund, med skruelåg
Flasker Plastflasker
PP, transparent.
  • Very good chemical resistance
  • Heat resistant up to +125 °C, autoclavable
  • Suitable for use with microwave ovens
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.