Beholdere, bred åbning, firkantede, med gevind
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
311-98311 311-83981
215-2576EA 127 DKK
215-2576 KAUT311-83981
Beholdere, bred åbning, firkantede, med gevind
Prøvecontainere Allround kasser
HDPE. Square. Natural colour. With moulded hand-grip.
  • Space-saving
  • Moulded hand-grip
  • Convenient handling
  • Suitable for food
  • Square shape saves space
  • Halal certified
Ideal for collecting, storing, and transporting solid chemicals. Nominal capacity 4400 ml not leakproof (wide neck container 4400 ml with screw closure).