Packaging bottles, narrow neck, round, LDPE, with screw caps, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312088-0004 312088-0016 312088-0032
NALG312088-0004EA 5260 DKK
NALG312088-0004 NALG312088-0016 NALG312088-0032
Packaging bottles, narrow neck, round, LDPE, with screw caps, Nalgene®
Flasker Plastflasker
LDPE, translucent, with PP screw cap.
  • Excellent impact and chemical resistance
  • Good temperature resistance
Designed for storage, shipping, and packaging of liquids.
Includes natural, polypropylene closures, ideal material for trace metal analysis because of LDPE low trace metal content, leakproof when used with Nalgene closures.
Certificeringer: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.