Koniske skilletragte, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
242941704 242942409 242943602 242944401 242945406 242946308 1064807 1064809 1064805 1064806
SCOT242941704EA 718 DKK
SCOT242941704 SCOT242942409 527-1217 SCOT242944401 SCOT242945406 SCOT242946308 SCOT1064807 SCOT1064809 SCOT1064805 SCOT1064806
Koniske skilletragte, DURAN®
Tragte Skilletragte
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.