Flasker, bred åbning, rektangulære, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312009-0004 312009-0008 312009-0016 312009-0032
NALG312009-0004EA 12940 DKK
NALG312009-0004 NALG312009-0008 NALG312009-0016 NALG312009-0032
Flasker, bred åbning, rektangulære, med skruelåg, Nalgene®
Flasker Plastflasker
Rectangular opaque amber bottles are made up of HDPE designed to protect light sensitive liquids or dry materials.
  • Space saving
  • Chemical- and puncture resistance
  • Leakproof sealing with opaque PP closures
  • Non sterile
Certificeringer: Compliance with ISO 13485:2003; 21CFR177.1520 and USP Class VI.
Leveringsinformation: Bottles are supplied with opaque amber PP closure.
Emballering: Bottles and closures are packed in separate polybags within the same carton.