Diagnostikflasker, smal åbning, runde, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312006-9050 312006-9125 312006-9025
NALG312006-9050EA 20080 DKK
NALG312006-9050 NALG312006-9125 NALG312006-9025
Diagnostikflasker, smal åbning, runde, med skruelåg, Nalgene®
Flasker Plastflasker
Transluscent bottles with closure used as sampling and to store and ship small volume and valuable reagents.
  • Nonsterile and durable
  • Linerless polypropylene closures that ensure leakproof performance
Certificeringer: Compliance with ISO 13485:2003 and 21CFR177.1520 USP Class VI.
Emballering: Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.