Mikro-filtreringstragte
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
07190410 07190310 07190010 07190110 07190210
LENZ07190410EA 276 DKK
LENZ07190410 LENZ07190310 LENZ07190010 LENZ07190110 LENZ07190210
Mikro-filtreringstragte
Tragte Filtreringstragte Mikrofiltertragte
Filter funnels, made of borosilicate glass 3,3, with integrated sintered glass filter plate.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance