Screw caps for laboratory bottles
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
07105500 07105501 07105502
LENZ07105500EA 823 DKK
LENZ07105500 LENZ07105501 LENZ07105502
Screw caps for laboratory bottles
Låg Flaskelåg
PBT, red, with PTFE coated seals.
  • For DIN GL threads
  • Heat resistant up to +180 °C
  • Autoclavable, sterilisable with hot air