HyperSep™ Hypercarb™ SPE columns
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60106-304 60106-403 60106-402 60106-303 60106-302 60106-301 60106-404
549-0624EA 3870 DKK
549-0624 549-0626 549-0625 549-0623 549-0622 549-0621 549-0627
HyperSep™ Hypercarb™ SPE columns
Kromatografikolonner
Flat, 100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents. Hydrophobic reversed phase; normal phase adsorption; polar retention.
  • Provides total pH stability and the retention and separation of highly polar species where silica and resin columns fail
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • No restriction on eluents