Kolber, rundbundede, smal åbning, med krave
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
07030028 07030037 07030049 07030058 07030070 07030073 07030076 07030079 07030082 07030085 07030088
LENZ07030028EA 1020 DKK
LENZ07030028 LENZ07030037 LENZ07030049 LENZ07030058 LENZ07030070 LENZ07030073 LENZ07030076 LENZ07030079 LENZ07030082 LENZ07030085 LENZ07030088
Kolber, rundbundede, smal åbning, med krave
Kolber Kolber med rund bund
DURAN® Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
Certificeringer: Acc. to DIN ISO 1773 and in addition to standard