Stophaner, laterale
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
81167 81248 81927
BRND81167EA 377 DKK
BRND81167 BRND81248 BRND81927
Stophaner, laterale
Stophaner
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.