Stophaner, nåleventil
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
82120 82121 82122 82170 82220 82221 82222
BRND82120EA 517 DKK
BRND82120 BRND82121 BRND82122 BRND82170 BRND82220 BRND82221 BRND82222
Stophaner, nåleventil
Stophaner
Borosilicate glass 3.3.
  • Interchangeable valve spindle
  • Spindle with retention device
  • Lateral or straight needle valve available
Valve housing with precision burette tip, PTFE spindle with PP cap.