HyperSep™ Retain™-AX SPE columns
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60107-404 60107-403 60107-401 60107-405 60107-406 60107-407 60107-408 60107-411 60107-412 60107-414 60107-415 60107-417 60107-419
549-0655EA 2870 DKK
549-0655 549-0654 549-0652 549-0656 549-0657 549-0658 549-0659 549-0660 549-0661 549-0662 549-0663 549-0886 549-0667
HyperSep™ Retain™-AX SPE columns
Kromatografikolonner
HyperSep™ Retain™-AX polymeric SPE material is a high purity, highly porous polystyrene DVB (divinyl benzene) material partially functionalised with quaternary amine groups to give balanced retention of acidic and non polar analytes. HyperSep™ Retain™-AX is ideal for the retention of THC and its metabolites.
  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14
HyperSep™ Retain™-AX offers reproducible and reliable sample preparation through excellent  batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™-AX. This can lead to fast, reproducible sample preparation and method development.