HyperSep™ Retain™ PEP SPE columns
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60107-203 60107-212 60107-204 60107-201 60107-206 60107-205 60107-207 60107-208 60107-211 60107-215 60107-217 60107-218
549-0630EA 1410 DKK
549-0630 549-0637 549-0631 549-0628 549-0633 549-0632 549-0634 549-0635 549-0636 549-0639 549-0664 549-0665
HyperSep™ Retain™ PEP SPE columns
Kromatografikolonner
HyperSep™ Retain™ PEP polymeric SPE material is a high purity, highly porous polystyrene DVB (divinyl benzene) material modified with urea functional groups to give balanced retention of polar and non polar analytes. HyperSep™ Retain™ PEP is ideal for a wide range of applications, such as drugs and metabolites in biological fluids, peptides in serum, plasma or biological fluids and environmental fluids.
  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14
HyperSep™ Retain™ PEP offers reproducible and reliable sample preparation through excellent  batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™ PEP. This can lead to fast sample preparation and method development.