Reagensglas med standardslib, med inddelinger
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05706001 05706002 05706003 05706004
LENZ05706001EA 179 DKK
LENZ05706001 LENZ05706002 LENZ05706003 LENZ05706004
Reagensglas med standardslib, med inddelinger
Rør Reagensglas
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round bottom glass with NS socket, with PE stopper, with scale in high-contrast enamel.