Reagensglas med standardslib, konisk bund
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05705015 05705016 05705115 05705116 05705216 05705215
LENZ05705015EA 82.6 DKK
LENZ05705015 LENZ05705016 LENZ05705115 LENZ05705116 LENZ05705216 LENZ05705215
Reagensglas med standardslib, konisk bund
Rør Reagensglas
Centrifuge tubes, made of DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes