Bottles, narrow neck, square, HDPE, with thread
Leverandør: Kautex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
310-85937 310-85938 310-85939 310-85940
KAUT310-85937EA 15.3 DKK
KAUT310-85937 215-6014 KAUT310-85939 KAUT310-85940
Bottles, narrow neck, square, HDPE, with thread
Flasker Plastflasker
HDPE, Natural colour.
  • Square shape
  • Safe and comfortable handling
  • Leakproof
  • Bottles are suitable for food
  • Halal certified
The neck of this bottle series has been adapted to the standard glass closure GL 32.