9,10-Dibromanthracen 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
112710100
112710500
ACRO112710100EA
472
DKK
ACRO112710100
ACRO112710500
9,10-Dibromanthracen 96%
9,10-Dibromoanthracene
Formel:
C₁₄H₈Br₂ Molekylvægt: 336.03 g/mol Kogepunkt: 427 Smeltepunkt: 220…225 °C |
MDL Number:
MFCD00001244 CAS-nummer: 523-27-3 EINECS: 208-342-4 Merck Index: 13,03041 |
Specifikationer Testresultater
Appearance | Yellow to yellow-green Powder |
Infrared spectrum | Conforms |
Melting point | 220 to 225 °C |
GC | ≥95 % |
Iodide (I) | ≤0.001 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Sodium (Na) | ≤0.005 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Nitrogen compounds (as N) | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...