1-Pentin ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 99%
CAS No.: 627-19-0
Molecular Formula: C5H8
Formula Weight: 68.12
Storage Temperature: 2°C to 8°C
Flash Point: -20°(-4°F)
Refractive Index: 1.3860
Odour: Unpleasant
Melting Point: -106°
Boiling Point: 39-41°
Beilstein Reference: 1697133
UN No.: 3295
MDL No.: MFCD00009469
Formel:
C₅H₈ Kogepunkt: 39…41 °C Smeltepunkt: –106 °C Massefylde: 0,692 Flammepunkt: –20 °C (–4 °F) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00009469 CAS-nummer: 627-19-0 EINECS: 210-987-1 UN nummer: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...