1-Pentin 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

303820250 303820050 303821000
ACRO303820250EA 750 DKK
ACRO303820250 ACRO303820050 ACRO303821000
1-Pentin 99%
1-Pentyne
Formel: C₅H₈
Kogepunkt: 40 °C
Smeltepunkt: 106…105 °C
Massefylde: 0,69 g/cm³
Flammepunkt: <–34 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009469
CAS-nummer: 627-19-0
EINECS: 210-987-1
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3840 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR