Oil sorbents
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HP156 HP100 T150 TF2001
MMMAHP156EA 2630 DKK
MMMAHP156 MMMAHP100 MMMAT150 MMMATF2001
Oil sorbents
Spild materialer
Oil adsorbent sheets or pads on a roll for spill control.
  • Hydrophobic
  • Oil (not water) is recovered
  • Can be used outside as well as in-plant