1-Penten ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A16723.14 A16723.22
A16723.14EA 517 DKK
A16723.14 A16723.22
1-Penten ≥97%
1-Pentene

Purity: 97% CAS No.: 109-67-1 Molecular Formula: CH3(CH2)2CH=CH2 Formula Weight: 70.14 Storage Temperature: 2°C to 8°C Flash Point: -51°(-60°F) Refractive Index: 1.3715 Melting Point: -138° Boiling Point: 30-31° Beilstein Reference: 1731629 Merck Reference: 14,7122 UN No.: 1108 MDL No.: MFCD00003567

Formel: C₅H₁₀
Kogepunkt: 30…31 °C
Smeltepunkt: –138 °C
Massefylde: 0,640
Flammepunkt: –51 °C (–60 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003567
CAS-nummer: 109-67-1
EINECS: 203-694-5
UN nummer: 1108
ADR: 3,I
Merck Index: 14,7122

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR