1-Decen ca. 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

111910010 111910025
ACRO111910010EA 503 DKK
ACRO111910010 ACRO111910025
1-Decen ca. 95%
1-Decene
Formel: C₁₀H₂₀
Molekylvægt: 140.27 g/mol
Kogepunkt: 169…171 °C
Smeltepunkt: –66 °C
Massefylde: 0.741 g/cm³
Flammepunkt: 49 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009577
CAS-nummer: 872-05-9
EINECS: 212-819-2
UN nummer: 3295
ADR: 3 III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥94 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR