Tape, polyimidfilm, 5413, 3M™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
54132533 5413633 54131233 54131933
MMMA54132533EA 2100 DKK
MMMA54132533 MMMA5413633 MMMA54131233 MMMA54131933
Tape, polyimidfilm, 5413, 3M™
Tape
Polyimide film tape consists of Kapton® polyimide film and silicon adhesive.
  • Dimensionally stable at high temperatures
  • Higher productivity by reducing rework
  • Flame retardant, chemical and radiation resistant
  • Temperature range: −73 to 260 °C
Applications include a mask for protection of gold fingers of printed circuit boards during wave solder or solder dip process or release surface in fabrication of parts cured at elevated temperatures.
Length: 33,0 m
Colour: Amber