Målekolber, klasse A
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
36220 36228 36238 36248 36254 129250 129252 36208
612-4918EA 1770 DKK
612-4918 612-4919 612-4920 612-4921 612-4923 612-4905 612-4907 612-4917
Målekolber, klasse A
Kolber Volumetriske kolber
PFA, semi-transparent, med PFA-skruelåg.
  • Inclusive one batch certificate
  • Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
Error limits according to class A, DIN EN ISO 1042. Calibrated to contain (TC, In). PFA screw cap with molded sealing ring. Autoclavable, easy to clean.
*Bred hals.
(Height = Height without stopper).