Målebægre med håndtag, PP
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7600275 7600280 7600286 7600290 7600293 7600294 7600297 7600296
HIRS7600275EA 500 DKK
HIRS7600275 HIRS7600280 HIRS7600286 HIRS7600290 HIRS7600293 HIRS7600294 HIRS7600297 HIRS7600296
Målebægre med håndtag, PP
Bægre
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
  • With spout
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products according to EC Directive No. 10/2011.