Målecylindre, lav form, klasse B, PP
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41620 41628 41638 41648 41662 41654
BRND41620EA 319 DKK
BRND41620 BRND41628 BRND41638 BRND41648 BRND41662 BRND41654
Målecylindre, lav form, klasse B, PP
Cylindere Målecylindre
PP, low form, Class B.
  • Calibrated to contain (TC, In)
  • Embossed scale
  • PP with high clarity
Robust cylinders which on exposure to temperatures up to 80 °C will not cause permanently exceeded tolerance/error limits.