Tragte
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
148215 148220 148225 148235 148237 148240 148245
BRND148215EA 838 DKK
BRND148215 BRND148220 BRND148225 BRND148235 BRND148237 BRND148240 BRND148245
Tragte
Tragte Filtreringstragte Filtertragte
PP, translucent.
  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers
For rapid filtration.