Hempelkolonner med vakuumkappe
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05119504 05119201 05119304 05119804
LENZ05119504EA 2090 DKK
LENZ05119504 LENZ05119201 LENZ05119304 LENZ05119804
Hempelkolonner med vakuumkappe
Destillations komponenter Destillationskolonner
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For packings like Raschig rings or glass beads, with NS socket and NS cone, with fixed vacuum jacket.