Buretterør uden stophaner
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3071260 3071270 3071275
HIRS3071260EA 239 DKK
HIRS3071260 HIRS3071270 HIRS3071275
Buretterør uden stophaner
Buretter
Burette tube, class B, DURAN® borosilicate glass, with blue graduation, calibrated for EX.
  • Head at bottom
  • Stopcock attachment, 8 mm Ø
  • Suitable for replaceable stopcock