Graduated pipettes, type 2, Class AS, total delivery
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1100360 1100329 1100365 1100354 1100353 1100342 1100341 1100335 1100333 1100370
612-4149EA 80.2 DKK
612-4149 HIRS1100329 612-4150 612-4148 HIRS1100353 612-4147 HIRS1100341 612-4146 612-4145 612-4152
Graduated pipettes, type 2, Class AS, total delivery
Pipetter Pipetter med inddeling
AR-Glas®.
  • Calibrated to deliver (TD, Ex); Type 2: nominal volume on top
  • Marks and inscriptions in ink durable brown
  • With standard cotton plug from five ml
  • Certificate of conformity with dated batch identification available on internet (www.hirschmannlab.de)
Length: 360 mm ±5 mm except for 25 pipettes: 450 mm ±5 mm
Certificeringer: DIN EN ISO 835. Conformity certified (DE-M marking).